Learn More
Selleck Chemical LLC A-438079 HCl 100mg 899431-18-6
SDP
Catalog No. 501365155
Encompass_Preferred
A-438079 HCl is a potent, and selective P2X7 receptor antagonist with pIC50 of 6.9. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5155 Supplier Selleck Chemical LLC Supplier No. S7705100MG
A-438079 HCl is a potent, and selective P2X7 receptor antagonist with pIC50 of 6.9. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.