Learn More
Medchemexpress LLC A 438079 hydrochloride | 899431-18-6 | C13H10Cl3N5 | 1 ML
SDP
Catalog No. 5000376073
Encompass_Preferred
A 438079 hydrochloride is a potent and selective P2X7 receptor antagonist with a pIC50 of 6.9, suitable for research applications. It blocks BzATP-evoked changes in intracellular calcium concentrations with an IC50 of 321 nM in rat P2X7 receptor-expressing cells, showing selectivity up to 100 μM. It has demonstrated efficacy in reducing pain and seizure severity in various in vivo models.
  • Potent and selective P2X7 receptor antagonist
  • Blocks BzATP-evoked intracellular calcium changes
  • Selective for P2X7 receptor
  • Reduces noxious and innocuous evoked activity in neuropathic rats
  • Raises withdrawal thresholds in SNL and CCI models
  • Reduces seizure severity and neuronal death
  • Prevents 6-OHDA-induced depletion of striatal dopamine stores
  • Reduces nociceptive behavior
Catalog No. 50-003-76073 Supplier Medchemexpress LLC Supplier No. HY15488A10MM/1ML
A 438079 hydrochloride is a potent and selective P2X7 receptor antagonist with a pIC50 of 6.9, suitable for research applications. It blocks BzATP-evoked changes in intracellular calcium concentrations with an IC50 of 321 nM in rat P2X7 receptor-expressing cells, showing selectivity up to 100 μM. It has demonstrated efficacy in reducing pain and seizure severity in various in vivo models.
  • Potent and selective P2X7 receptor antagonist
  • Blocks BzATP-evoked intracellular calcium changes
  • Selective for P2X7 receptor
  • Reduces noxious and innocuous evoked activity in neuropathic rats
  • Raises withdrawal thresholds in SNL and CCI models
  • Reduces seizure severity and neuronal death
  • Prevents 6-OHDA-induced depletion of striatal dopamine stores
  • Reduces nociceptive behavior