Learn More
Medchemexpress LLC A 438079 hydrochloride | 899431-18-6 | C13H10Cl3N5 | 5 MG
SDP
Catalog No. 5000376487
Encompass_Preferred
A 438079 hydrochloride is a potent and selective P2X7 receptor antagonist with a pIC50 of 6.9. It is intended for research use only and not for patient administration.
  • Potent and selective P2X7 receptor antagonist
  • High purity of 99.99%
  • Appearance as a solid, white to off-white
  • For research use only
Catalog No. 50-003-76487 Supplier Medchemexpress LLC Supplier No. HY15488A5MG
A 438079 hydrochloride is a potent and selective P2X7 receptor antagonist with a pIC50 of 6.9. It is intended for research use only and not for patient administration.
  • Potent and selective P2X7 receptor antagonist
  • High purity of 99.99%
  • Appearance as a solid, white to off-white
  • For research use only