Learn More
Cayman Chemical A-438079hydrochlorIde 10mg
SDP
Catalog No. 502968948
Encompass_Preferred
A competitive antagonist of the nucleotide receptor P2X7 (pIC50 = 6.9); inhibits both calcium flux and IL-1β release mediated by P2X7 while not affecting the activity of other P2X receptors; used to evaluate the role of P2X7 in nociception, oxidative stress, and apoptosis in cells and animals
Catalog No. 50-296-8948 Supplier Cayman Chemical Supplier No. 1458010 MG
A competitive antagonist of the nucleotide receptor P2X7 (pIC50 = 6.9); inhibits both calcium flux and IL-1β release mediated by P2X7 while not affecting the activity of other P2X receptors; used to evaluate the role of P2X7 in nociception, oxidative stress, and apoptosis in cells and animals