Learn More
Selleck Chemical LLC A-443654-E1125-5MG
SDP
Catalog No. 503134664
Encompass_Preferred
A-443654 a derivative of indazole pyridine compounds is a pan Akt (Akt1 2 3 isoforms) inhibitor which binds to the ATP-binding site of Akt It is an ATP competitive and reversible inhibitor
Catalog No. 50-313-4664 Supplier Selleck Chemical LLC Supplier No. E11255MG
A-443654 a derivative of indazole pyridine compounds is a pan Akt (Akt1 2 3 isoforms) inhibitor which binds to the ATP-binding site of Akt It is an ATP competitive and reversible inhibitor