Learn More
Medchemexpress LLC A-484954 | 142557-61-7 | 99.6% | 289.29 | 10 MG
SDP
Catalog No. 5000344906
Encompass_Preferred
A-484954 is a highly selective eukaryotic elongation factor-2 kinase (eEF2K) inhibitor with an IC50 of 280 nM. It is intended for research use only.
  • Highly selective eEF2K inhibitor
  • Inhibits MDA-MB-231 cell viability at IC50 99480 nM
  • Causes relaxation in E (+) and E (-) aorta or mesenteric artery
  • Inhibits MCT-induced increases in PA pressure, PA hypertrophy, and fibrosis
Catalog No. 50-003-44906 Supplier Medchemexpress LLC Supplier No. HY1100961MG
A-484954 is a highly selective eukaryotic elongation factor-2 kinase (eEF2K) inhibitor with an IC50 of 280 nM. It is intended for research use only.
  • Highly selective eEF2K inhibitor
  • Inhibits MDA-MB-231 cell viability at IC50 99480 nM
  • Causes relaxation in E (+) and E (-) aorta or mesenteric artery
  • Inhibits MCT-induced increases in PA pressure, PA hypertrophy, and fibrosis