Learn More
Medchemexpress LLC HY-107455 100mg , A-485 CAS:1889279-16-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107455100MG
Encompass_Preferred
HY-107455 100mg A-485 CAS: 1889279-16-6 A-485 is a potent and selective catalytic inhibitor of p300/CBP with IC50s of 9.8 nM and 2.6 nM for p300 and CBP histone acetyltransferase (HAT), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9308