Learn More
Miele, Inc. A 500
Catalog No. 501953757
Encompass_Preferred
A 500 Load carrierSingle-rack load carrier, extendable to 2 levelsFor inserts (and possibly mesh trays) and for cleaning laboratory glassware and utensilsExtendable using A 603 (not included)H 153, W 650, D 790 mm
Catalog No. 50-195-3757 Supplier Miele, Inc. Supplier No. 10973880
A 500 Load carrierSingle-rack load carrier, extendable to 2 levelsFor inserts (and possibly mesh trays) and for cleaning laboratory glassware and utensilsExtendable using A 603 (not included)H 153, W 650, D 790 mm