Learn More
Miele, Inc. A 501 Four-level mobile unit f
Catalog No. 502870702
Encompass_Preferred
A 501 Four-level mobile unit for inserts and cleaning glasswares, with 3 spray arms. Extendable to 5-6 levels with a separate A601
Catalog No. 50-287-0702 Supplier Miele, Inc. Supplier No. 12684440
Add to Cart
Add to Cart
Request a Quote
A 501 Four-level mobile unit for inserts and cleaning glasswares, with 3 spray arms. Extendable to 5-6 levels with a separate A601