Learn More
Miele, Inc. A 501
Catalog No. 501953758
Encompass_Preferred
A 501 Load carrierLoad carrier with 4 levels, extendable to 5 or 6 levelsFor inserts (and possibly mesh trays) and for cleaning laboratory glassware and utensilsMatching insertable rack level A 601 (not included)H 659, W 640, D 790 mm
Catalog No. 50-195-3758 Supplier Miele, Inc. Supplier No. 10973890
Add to Cart
Add to Cart
Request a Quote
A 501 Load carrierLoad carrier with 4 levels, extendable to 5 or 6 levelsFor inserts (and possibly mesh trays) and for cleaning laboratory glassware and utensilsMatching insertable rack level A 601 (not included)H 659, W 640, D 790 mm