Learn More
Miele, Inc. A 503 Mobile unit for inserts
Catalog No. 502870725
Encompass_Preferred
A 503 Mobile unit for inserts and cleaning glasswares, up to 6 injector modules on 2 levels
Catalog No. 50-287-0725 Supplier Miele, Inc. Supplier No. 12685380
A 503 Mobile unit for inserts and cleaning glasswares, up to 6 injector modules on 2 levels