Learn More
Miele, Inc. A 505
Catalog No. 501953760
Encompass_Preferred
A 505 Load carrierThe A 505 load carrier can accommodate inserts or various mesh trays and laboratory utensils.H 524, W 640, D 790 mmLoading dimensionsLevel 1: H 202, W 585, D 780 mmLevel 2: H 202, W 590, D 780 mmLevel 3: H 132, W 590, D 780 mm
Catalog No. 50-195-3760 Supplier Miele, Inc. Supplier No. 10998970
A 505 Load carrierThe A 505 load carrier can accommodate inserts or various mesh trays and laboratory utensils.H 524, W 640, D 790 mmLoading dimensionsLevel 1: H 202, W 585, D 780 mmLevel 2: H 202, W 590, D 780 mmLevel 3: H 132, W 590, D 780 mm