Learn More
Miele, Inc. A 506
Catalog No. 501953761
Encompass_Preferred
A 506 Load carrierThe A 506 load carrier can accommodate inserts or various mesh trays for cleaning laboratory glassware and utensils.H 557, W 640, D 790 mmLoading dimensionsLevel 1: H 112.5, W 585, D 780 mmLevel 2: H 112.5, W 595, D 780 mmLevel 3: H 112.5, W 595, D 780 mmLevel 4: H 114.0, W 595, D 780 mm
Catalog No. 50-195-3761 Supplier Miele, Inc. Supplier No. 10999130
Add to Cart
Add to Cart
Request a Quote
A 506 Load carrierThe A 506 load carrier can accommodate inserts or various mesh trays for cleaning laboratory glassware and utensils.H 557, W 640, D 790 mmLoading dimensionsLevel 1: H 112.5, W 585, D 780 mmLevel 2: H 112.5, W 595, D 780 mmLevel 3: H 112.5, W 595, D 780 mmLevel 4: H 114.0, W 595, D 780 mm