Learn More
Cayman Chemical A-54556B 2.5mg
SDP
Catalog No. 502967980
Encompass_Preferred
A natural acyldepsipeptide that is effective against Gram-positive bacteria, including penicillin-resistant S. aureus; is also effective against many strains of anaerobic bacteria
Catalog No. 50-296-7980 Supplier Cayman Chemical Supplier No. 170902.5 MG
A natural acyldepsipeptide that is effective against Gram-positive bacteria, including penicillin-resistant S. aureus; is also effective against many strains of anaerobic bacteria