Learn More
Lauda Brinkmann Lp A 6 HEATING CIRCULATOR
Catalog No. 50931208
Encompass_Preferred
Heating Circulator, Alpha Series, Model A 6, Digital control interface with simple 3 button operation, 6 L bath size, 1.15 kW Heater, Temperature range of ambient to 85 deg.C, Flow rate from 5 to 15 L/min., 115V; 60Hz This item is non returnable*
Catalog No. 50-931-208 Supplier Lauda Brinkmann Lp Supplier No. L000631
Heating Circulator, Alpha Series, Model A 6, Digital control interface with simple 3 button operation, 6 L bath size, 1.15 kW Heater, Temperature range of ambient to 85 deg.C, Flow rate from 5 to 15 L/min., 115V; 60Hz This item is non returnable*