Learn More
Miele, Inc. A 603
Catalog No. 501953763
Encompass_Preferred
A 603 Rack levelFor use on A 500, with spray armThe A 603 module can be used to add a rack level with a spray arm to the A 500 load carrier.W 611, H 372, D 784 mm
Catalog No. 50-195-3763 Supplier Miele, Inc. Supplier No. 10973980
A 603 Rack levelFor use on A 500, with spray armThe A 603 module can be used to add a rack level with a spray arm to the A 500 load carrier.W 611, H 372, D 784 mm