Learn More
Miele, Inc. A 605
Catalog No. 501953764
Encompass_Preferred
A 605 ModuleThe module can be used on load carrier A 503.H 160, W 255, D 611 mm. The module is suitable for items with a maximum diameter of 20 mm and a maximum height of 160 mm.
Catalog No. 50-195-3764 Supplier Miele, Inc. Supplier No. 10999010
A 605 ModuleThe module can be used on load carrier A 503.H 160, W 255, D 611 mm. The module is suitable for items with a maximum diameter of 20 mm and a maximum height of 160 mm.