Learn More
Miele, Inc. A 612
Catalog No. 501953766
Encompass_Preferred
A 612 ModuleThe A 612 injector module is suitable for reprocessing large-volume laboratory glassware. The module can be used on load carrier A 503.H 361, W 255, D 614 mmThe maximum diameter of the items depends on the number of nozzles used. Requires A 846, A 847 and/or A 848, depending on load.
Catalog No. 50-195-3766 Supplier Miele, Inc. Supplier No. 10999050
A 612 ModuleThe A 612 injector module is suitable for reprocessing large-volume laboratory glassware. The module can be used on load carrier A 503.H 361, W 255, D 614 mmThe maximum diameter of the items depends on the number of nozzles used. Requires A 846, A 847 and/or A 848, depending on load.