Learn More
Miele, Inc. A 613
Catalog No. 501953770
Encompass
A 613 ModuleThe A 613 module is designed for reprocessing volumetric pipettes and measuring pipettes. The module can be used on load carrier A 503.H 195, W 255, D 614 mm
Catalog No. 50-195-3770 Supplier Miele, Inc. Supplier No. 10999070
A 613 ModuleThe A 613 module is designed for reprocessing volumetric pipettes and measuring pipettes. The module can be used on load carrier A 503.H 195, W 255, D 614 mm