Learn More
Apexbio Technology LLC A 61603 hydrobromide 107756-30-9 10mg
SDP
Catalog No. 5000092038
Encompass_Preferred
A 61603 hydrobromide (CAS 107756-30-9) is a potent and highly selective agonist of the 1A-adrenergic receptor one of the 1-adrenergic receptor subtypes involved in cellular growth and proliferation Compared to its activity at 1B- and 1D-adrenergic receptors A 61603 demonstrates approximately 35-fold higher potency for 1A-AR In cellular assays it robustly induces phosphoinositide hydrolysis in 1A-AR-expressing fibroblasts (pEC50 8 24) and in neonatal rat cardiomyocytes it increases calcium transient frequency in a dose-dependent manner (EC50 6 9 nM) In vascular models A 61603 markedly elevates perfusion pressure demonstrating significantly greater efficacy than noradrenaline This compound serves as a valuable tool for delineating 1A-AR-mediated signaling and vascular regulation
Catalog No. 50-000-92038 Supplier Apexbio Technology LLC Supplier No. B657110
A 61603 hydrobromide (CAS 107756-30-9) is a potent and highly selective agonist of the 1A-adrenergic receptor one of the 1-adrenergic receptor subtypes involved in cellular growth and proliferation Compared to its activity at 1B- and 1D-adrenergic receptors A 61603 demonstrates approximately 35-fold higher potency for 1A-AR In cellular assays it robustly induces phosphoinositide hydrolysis in 1A-AR-expressing fibroblasts (pEC50 8 24) and in neonatal rat cardiomyocytes it increases calcium transient frequency in a dose-dependent manner (EC50 6 9 nM) In vascular models A 61603 markedly elevates perfusion pressure demonstrating significantly greater efficacy than noradrenaline This compound serves as a valuable tool for delineating 1A-AR-mediated signaling and vascular regulation