Learn More
Miele, Inc. A 621
Catalog No. 501953772
Encompass_Preferred
A 621 ModuleModule, empty, with 20 injector positionsFor cleaning narrow-necked laboratory glasswareFor use in A 503H 113, W 184, D 614 mm
Catalog No. 50-195-3772 Supplier Miele, Inc. Supplier No. 10999110
A 621 ModuleModule, empty, with 20 injector positionsFor cleaning narrow-necked laboratory glasswareFor use in A 503H 113, W 184, D 614 mm