Learn More
Miele, Inc. A 622 Module 4x9 SimpleLoad
Catalog No. 502870795
Encompass_Preferred
A 622 Module 4x9, SimpleLoad, for use in A 503, 36 empty positions
Catalog No. 50-287-0795 Supplier Miele, Inc. Supplier No. 12678490
A 622 Module 4x9, SimpleLoad, for use in A 503, 36 empty positions