Learn More
Miele, Inc. A 622
Catalog No. 501953773
Encompass_Preferred
A 622 ModuleModule, empty, with 40 injector positionsFor cleaning narrow-necked laboratory glasswareFor use in A 503H 113, W 208, D 614 mm
Catalog No. 50-195-3773 Supplier Miele, Inc. Supplier No. 10999120
A 622 ModuleModule, empty, with 40 injector positionsFor cleaning narrow-necked laboratory glasswareFor use in A 503H 113, W 208, D 614 mm