Learn More
Eurofins Discoverx A-674563, 2 mg. (92-1244)
Supplier:  Eurofins Discoverx 921244
Encompass
A-674563 is a synthetic small molecule inhibitor of Akt1, as well as PKA and CDK2 (IC50=11, 16, and 46 nM, respectively). >30-fold selective over PKC. Provided as 2mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9765