Learn More
TARGETMOL CHEMICALS INC A-674563 2HCL552325-73-2 5MG
SDP
Catalog No. 502579521
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A-674563 2HCl is an Akt1 inhibitor with Ki of 11 nM modest potent to PKA and >30-fold selective for Akt1 over PKC.
Catalog No. 50-257-9521 Supplier TARGETMOL CHEMICALS INC Supplier No. T6139L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A-674563 2HCl is an Akt1 inhibitor with Ki of 11 nM modest potent to PKA and >30-fold selective for Akt1 over PKC.