Learn More
AdipoGen A-674563 free base
SDP
Catalog No. 502562234
Encompass_Preferred
Chemical. CAS 552325-73-2. Formula C22H22N4O. MW 358.4. Synthetic. A-674563 is a potent, ATP-competitive, reversible and orally available Akt1 inhibitor with an IC50 of 11nM. Binds to the ATP site of the AKT kinase domain, inhibiting AKT-catalyzed phosphorylation activity. Also inhibits PKA and CDK2 with IC50 values of 16nM and 46nM, respectively. 10- to >1,800-fold selective for Akt1 versus additional kinases in the CMGC, CAMK and TK families. Anticancer compound. Reduces phosphorylation of Akt downstream targets in cells and slows proliferation of tumor cells in vitro with an EC50 value of 0.4µM. Shown to decrease tumor growth in mouse tumor xenograft models. Decreased sphingosine kinase 1 SphK1 activity in AML cells to deplete pro-survival sphingosine-1-phosphate S1P and boost pro-apoptotic ceramide production.
Catalog No. 50-256-2234 Supplier AdipoGen Supplier No. AGCR13680M010
Chemical. CAS 552325-73-2. Formula C22H22N4O. MW 358.4. Synthetic. A-674563 is a potent, ATP-competitive, reversible and orally available Akt1 inhibitor with an IC50 of 11nM. Binds to the ATP site of the AKT kinase domain, inhibiting AKT-catalyzed phosphorylation activity. Also inhibits PKA and CDK2 with IC50 values of 16nM and 46nM, respectively. 10- to >1,800-fold selective for Akt1 versus additional kinases in the CMGC, CAMK and TK families. Anticancer compound. Reduces phosphorylation of Akt downstream targets in cells and slows proliferation of tumor cells in vitro with an EC50 value of 0.4µM. Shown to decrease tumor growth in mouse tumor xenograft models. Decreased sphingosine kinase 1 SphK1 activity in AML cells to deplete pro-survival sphingosine-1-phosphate S1P and boost pro-apoptotic ceramide production.