Learn More
Medchemexpress LLC A-674563 hydrochloride | 2070009-66-2 | 99.6% | 394.90 | 5 MG
SDP
Catalog No. 5000364631
Encompass_Preferred
A-674563 hydrochloride is a potent and selective Akt1 inhibitor with a Ki of 11 nM. This product is suitable for laboratory research and includes the following features:
  • Potent and selective Akt1 inhibitor
  • Light yellow to brown solid appearance
  • Soluble in water and DMSO
  • Stored under nitrogen, away from moisture
Catalog No. 50-003-64631 Supplier Medchemexpress LLC Supplier No. 5MGHY13254A
A-674563 hydrochloride is a potent and selective Akt1 inhibitor with a Ki of 11 nM. This product is suitable for laboratory research and includes the following features:
  • Potent and selective Akt1 inhibitor
  • Light yellow to brown solid appearance
  • Soluble in water and DMSO
  • Stored under nitrogen, away from moisture