Learn More
TARGETMOL CHEMICALS INC A-69412 5MG
Supplier: TARGETMOL CHEMICALS INC T102085MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A-69412 (N-1-(Fur-3-ylethyl)-N-hydroxyurea) is a reversible specific inhibitor of the hydrophilic 5-lipoxygenase. It has the potential to treat ulcerative colitis and asthma and possibly other inflammatory and allergic conditions. purity: 99%