Learn More
Medchemexpress LLC HY-50697 100mg Medchemexpress, A-740003 CAS:861393-28-4 Purity:98%
SDP
Catalog No. 501639014
Encompass_Preferred
Medchemexpress,HY-50697 100mg A-740003 CAS:861393-28-4 Formula:C26H30N6O3 A-740003 is a potent, selective and competitive P2X7 receptor antagonist with IC50 values are 18 and 40 nM for rat and human P2X7 receptors, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9014 Supplier Medchemexpress LLC Supplier No. HY50697100MG
Medchemexpress,HY-50697 100mg A-740003 CAS:861393-28-4 Formula:C26H30N6O3 A-740003 is a potent, selective and competitive P2X7 receptor antagonist with IC50 values are 18 and 40 nM for rat and human P2X7 receptors, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.