Learn More
Medchemexpress LLC HY-50662 10mg Medchemexpress, A-769662 CAS:844499-71-4 Purity:>98%
SDP
Catalog No. 502029323
Encompass_Preferred
Medchemexpress, HY-50662 10mg A-769662 CAS:844499-71-4 A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9323 Supplier Medchemexpress LLC Supplier No. HY5066210MG
Medchemexpress, HY-50662 10mg A-769662 CAS:844499-71-4 A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.