Learn More
Apexbio Technology LLC A-769662, 844499-71-4, MFCD11977269, 25 mg
SDP
Supplier:  Apexbio Technology LLC A396325
Encompass_Preferred
MF: C20H12N2O3S, MW: 360.39, Solubility: 18.0195 mg/mL in DMSO, insoluble in EtOH, insoluble in H2O. A-769662 is a potent activator of AMPK with EC50 value of 0.8 uM in vitro.
Catalog No. 50-101-3719