Learn More
Medchemexpress LLC HY-50662 50mg , A-769662 CAS:844499-71-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5066250MG
Encompass_Preferred
HY-50662 50mg A-769662 CAS: 844499-71-4 A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1074