Learn More
Selleck Chemical LLC A-769662 5mg 844499-71-4
SDP
Supplier:  Selleck Chemical LLC S26975MG
Encompass_Preferred
A-769662 is a potent, reversible AMPK activator with EC50 of 0.8 ?M in cell-free assays, little effect on GPPase/FBPase activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2795