Learn More
Abcam A 769662, AMP-activated protein kinase activator, 50MG
Catalog No. 502584623
Encompass_Preferred
MW 360.4 Da, Purity >99%. Potent, reversible activator of AMP-activated protein kinase (AMPK) (EC₅₀ = 0.8 μM). Directly activates native rat AMPK by mimicking the effects of AMP - it activates AMPK both allosterically and by inhibiting dephosphorylation of AMPK. Shown to inhibit Na+-K+-ATPase at higher concentrations (IC₅₀ = 57 μM) . Inhibits differentiation of adipocytes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4623 Supplier Abcam Supplier No. AB12033550MG
MW 360.4 Da, Purity >99%. Potent, reversible activator of AMP-activated protein kinase (AMPK) (EC₅₀ = 0.8 μM). Directly activates native rat AMPK by mimicking the effects of AMP - it activates AMPK both allosterically and by inhibiting dephosphorylation of AMPK. Shown to inhibit Na+-K+-ATPase at higher concentrations (IC₅₀ = 57 μM) . Inhibits differentiation of adipocytes.
The product is subject to the following: Abcam Restricted Use Statement