Learn More
Medchemexpress LLC A 77-01 | 607737-87-1 | 99.92% | C18H14N4 | 250 MG
SDP
Catalog No. 5000358612
Encompass_Preferred
A 77-01 is a potent inhibitor of the transforming growth factor (TGF)-β type I receptor superfamily activin-like kinase ALK5, with an IC50 of 25 nM. It is for research use only.
  • Potent inhibitor of ALK5 with an IC50 of 25 nM.
  • Effectively inhibits TGF-β-induced transcriptional activation.
  • Prevents the growth-inhibitory effects of TGF-β.
  • Inhibits TGF-β-induced phosphorylation of Smad2.
Catalog No. 50-003-58612 Supplier Medchemexpress LLC Supplier No. 250MGHY78349
A 77-01 is a potent inhibitor of the transforming growth factor (TGF)-β type I receptor superfamily activin-like kinase ALK5, with an IC50 of 25 nM. It is for research use only.
  • Potent inhibitor of ALK5 with an IC50 of 25 nM.
  • Effectively inhibits TGF-β-induced transcriptional activation.
  • Prevents the growth-inhibitory effects of TGF-β.
  • Inhibits TGF-β-induced phosphorylation of Smad2.