Learn More
Medchemexpress LLC A-77636 hydrochloride | 145307-34-2 | MFCD03107804 | ≥98.0% | 365.89 | C20H28ClNO3 | 5 MG
SDP
Catalog No. 5000244774
Encompass_Preferred
A-77636 hydrochloride is a potent, orally active, selective, and long-acting dopamine D1 receptor agonist used in pharmacological research, including studies of dopamine D1-like receptor signaling and antiparkinsonian activity.
  • Potent dopamine D1 receptor agonist (Ki ≈ 39.8 nM).
  • Selective for D1-like receptors with minimal D2 activity.
  • High purity (≥98.0%) suitable for research applications.
  • Solid powder form for ease of formulation and dosing.
  • Small 5 mg quantity useful for screening and early-stage experiments.
Catalog No. 50-002-44774 Supplier Medchemexpress LLC Supplier No. HY1034165MG
A-77636 hydrochloride is a potent, orally active, selective, and long-acting dopamine D1 receptor agonist used in pharmacological research, including studies of dopamine D1-like receptor signaling and antiparkinsonian activity.
  • Potent dopamine D1 receptor agonist (Ki ≈ 39.8 nM).
  • Selective for D1-like receptors with minimal D2 activity.
  • High purity (≥98.0%) suitable for research applications.
  • Solid powder form for ease of formulation and dosing.
  • Small 5 mg quantity useful for screening and early-stage experiments.