Learn More
TARGETMOL CHEMICALS INC A 77636 HYDROCHLORIDE 5MG
SDP
Catalog No. 502581004
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A 77636 hydrochloride is a potent orally active selective and long acting dopamine D1 receptor agonist (pKi=7.40; Ki=39.8 nM). A 77636 hydrochloride shows antiparkinsonian activity. A 77636 hydrochloride is functionally inactive at dopamine D2 receptor. purity: 99%
Catalog No. 50-258-1004 Supplier TARGETMOL CHEMICALS INC Supplier No. T217825MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A 77636 hydrochloride is a potent orally active selective and long acting dopamine D1 receptor agonist (pKi=7.40; Ki=39.8 nM). A 77636 hydrochloride shows antiparkinsonian activity. A 77636 hydrochloride is functionally inactive at dopamine D2 receptor. purity: 99%