Learn More
Medchemexpress LLC HY-P0216 1mg Medchemexpress, A 779 CAS:159432-28-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02161MG
Encompass_Preferred
Medchemexpress, HY-P0216 1mg A 779 CAS:159432-28-7 A 779 is a specific antagonist of G-protein coupled receptor (Mas receptor), which is an Ang1-7 receptor distinct from the classical AngII. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4055