Learn More
Selleck Chemical LLC A-779-E0039-25MG
SDP
Catalog No. 503135717
Encompass_Preferred
A-779 is a potent and selective antagonist for Angiotensin-(1-7) (Ang-(1-7)) with an IC50 of 0 3 nM in a radioligand binding assay A-779 shows no significant affinity for AT1 or AT2 receptors at a concentration of 1 M
Catalog No. 50-313-5717 Supplier Selleck Chemical LLC Supplier No. E003925MG
A-779 is a potent and selective antagonist for Angiotensin-(1-7) (Ang-(1-7)) with an IC50 of 0 3 nM in a radioligand binding assay A-779 shows no significant affinity for AT1 or AT2 receptors at a concentration of 1 M