Learn More
TARGETMOL CHEMICALS INC A 779 TFA159432-28-7 FRE 25MG
SDP
Catalog No. 502579173
Encompass_Preferred
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A 779 TFA is a potent antagonist of angiotensin-(1-7) receptor.
Catalog No. 50-257-9173 Supplier TARGETMOL CHEMICALS INC Supplier No. T7616L25MG
Add to Cart
Add to Cart
Also available in 1 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A 779 TFA is a potent antagonist of angiotensin-(1-7) receptor.