Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC A-802715 | 107767-58-8 | 322.40 | 1 ML
SDP

Catalog No. 5000448495
Encompass_Preferred

A-802715 is a methylxanthine derivative. It has a reported TD50 (toxic dose of 50%) of 0.9-1.1 mM. This product is for research use only.

  • Purity of 98.45%
  • Molecular weight of 322.40
  • CAS number 107767-58-8

Catalog No. 50-004-48495 Supplier Medchemexpress LLC Supplier No. HYU0014210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

A-802715 is a methylxanthine derivative. It has a reported TD50 (toxic dose of 50%) of 0.9-1.1 mM. This product is for research use only.

  • Purity of 98.45%
  • Molecular weight of 322.40
  • CAS number 107767-58-8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.