Learn More
Apexbio Technology LLC A-803467 944261-79-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063454
Encompass_Preferred
A-803467 (CAS 944261-79-4) is a potent selective blocker of the voltage-gated sodium channel subtype NaV1 8 It inhibits human and rat recombinant NaV1 8 channels with IC50 values of approximately 79 nM and 45 nM respectively Its selectivity for NaV1 8 over other sodium channel subtypes (e g NaV1 2 NaV1 3 NaV1 5 and NaV1 7) is 300 1000 fold By specifically reducing activity in tetrodotoxin-resistant sodium currents of dorsal root ganglion neurons A-803467 suppresses induced and spontaneous action potential firing exhibiting analgesic properties in animal models of neuropathic and inflammatory pain
Catalog No. 50-000-63454 Supplier Apexbio Technology LLC Supplier No. B22775.1
A-803467 (CAS 944261-79-4) is a potent selective blocker of the voltage-gated sodium channel subtype NaV1 8 It inhibits human and rat recombinant NaV1 8 channels with IC50 values of approximately 79 nM and 45 nM respectively Its selectivity for NaV1 8 over other sodium channel subtypes (e g NaV1 2 NaV1 3 NaV1 5 and NaV1 7) is 300 1000 fold By specifically reducing activity in tetrodotoxin-resistant sodium currents of dorsal root ganglion neurons A-803467 suppresses induced and spontaneous action potential firing exhibiting analgesic properties in animal models of neuropathic and inflammatory pain