Learn More
Apexbio Technology LLC A-803467, 50mg. Cas: 944261-79-4 MFCD: MFCD10574689. NaV1.8 channel blocker,potent and selective
SDP
Supplier:  Apexbio Technology LLC B227750
Encompass_Preferred
A-803467 is a potent and selective blocker of Nav1.8 sodium channel with IC50 value of 8nM [1].A-803467 is a sodium channel blocker with high affinity and selectivity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5338