Learn More
Enzo Life Sciences A-803467 (50mg). CAS: 944261-79-4
SDP
Supplier:  Enzo Life Sciences BMLNA1420050
Encompass_Preferred
A-803467 is a potent and selective inhibitor of the TTX-resistant voltage-gated sodium channel Nav1.8 which is preferentially expressed on sensory neurons. IC50 for human Nav1.8=8.0 nM (140 nM for rat). It attenuates neuropathic and inflammatory pain in various animal models. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (15mg/ml) or 100% ethanol (2mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1186