Learn More
Medchemexpress LLC HY-100483 5mg Medchemexpress, A-804598 CAS:1125758-85-1 Purity:>98%
SDP
Catalog No. 501931849
Encompass_Preferred
Medchemexpress, HY-100483 5mg A-804598 CAS:1125758-85-1 A-804598 is a CNS penetrant, competitive and selective P2X7 receptor antagonist with IC 50 s of 9 nM, 10 nM and 11 nM for mouse, rat and human P2X7 receptors, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1849 Supplier Medchemexpress LLC Supplier No. HY1004835MG
Medchemexpress, HY-100483 5mg A-804598 CAS:1125758-85-1 A-804598 is a CNS penetrant, competitive and selective P2X7 receptor antagonist with IC 50 s of 9 nM, 10 nM and 11 nM for mouse, rat and human P2X7 receptors, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.