Learn More
Apexbio Technology LLC A 83-01, 909910-43-6, MFCD08705403, 10 mg
SDP
Supplier:  Apexbio Technology LLC A313310
Encompass_Preferred
MF: C25H19N5S, MW: 421.52, Solubility: 21.1 mg/mL in DMSO, 9.82 mg/mL in EtOH with ultrasonic and warming, insoluble in H2O, Synonyms: A83-01; A-83-01. A 83-01 is a small-molecule inhibitor of activin receptorlike kinase (ALK)-5 with IC50 value of 12 nM.
Catalog No. 50-115-0476