Learn More
Medchemexpress LLC HY-10432 50mg Medchemexpress, A 83-01 CAS:909910-43-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1043250MG
Encompass_Preferred
Medchemexpress, HY-10432 50mg A 83-01 CAS:909910-43-6 A 83-01 is a potent inhibitor of TGF-β type I receptor ALK5 kinase, type I nodal receptor ALK4 and type I nodal receptor ALK7, with IC50s of 12, 45 and 7.5 nM against the transcription induced by ALK5, ALK4 and ALK7, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8217