Learn More
TARGETMOL CHEMICALS INC A 83-01 50MG
SDP
Catalog No. 502580419
Encompass_Preferred
Also available in 5 mg 10 mg and bulk. Please contact Fisher for quotes. A 83-01 (ALK5 Inhibitor IV) is a selective inhibitor of TGF-(beta) type I receptor ALK5 kinase type I activin/nodal receptor ALK4 and type I nodal receptor ALK7 with IC50 values of 12 nM 45 nM and 7.5 nM respectively. purity: 98%
Catalog No. 50-258-0419 Supplier TARGETMOL CHEMICALS INC Supplier No. T303150MG
Also available in 5 mg 10 mg and bulk. Please contact Fisher for quotes. A 83-01 (ALK5 Inhibitor IV) is a selective inhibitor of TGF-(beta) type I receptor ALK5 kinase type I activin/nodal receptor ALK4 and type I nodal receptor ALK7 with IC50 values of 12 nM 45 nM and 7.5 nM respectively. purity: 98%