Learn More
Apexbio Technology LLC A 83-01 909910-43-6 50mg
SDP
Supplier:  Apexbio Technology LLC A313350
Encompass_Preferred
A 83-01 (CAS 909910-43-6) is a selective small-molecule inhibitor targeting activin receptor-like kinase 5 (ALK-5) with additional inhibitory effects on ALK-4 and ALK-7 It specifically blocks ALK-5-mediated signalling induced by TGF- suppressing downstream Smad-dependent transcription with an IC50 of approximately 12 nM Assays in Mv1LuR4-2 cells showed that at 1 M A 83-01 reduced ALK-5-induced luciferase reporter activity by 68% By targeting these ALK receptors A 83-01 is frequently utilized to investigate TGF- signaling pathways epithelial-mesenchymal transition and cellular growth inhibition in biological research
Catalog No. 50-000-61418