Learn More
STEMCELL Technologies A 83-01, Size: 10 mg
SDP
Catalog No. 501975873
Encompass
Inhibit activin receptor-like kinase (ALK), including ALK5 (type I transforming growth factor-β receptor), ALK4 (type IB activin receptor), and ALK7 (type I NODAL receptor) with A 83-01 (Tojo et al. Cancer Sci, 2005). A 83-01 inhibits mesoderm and endoderm specification (Zhang et al. Cell Stem Cell, 2016), facilitates the conversion of epiblast stem cells to the naїve pluripotent state (Zhou et al. J Biol Chem, 2010; Li et al. Cell Stem Cell, 2009), and disrupts epithelial to mesenchymal transition through inhibition of the SMAD signaling pathway (Tojo et al. Cancer Sci, 2005). In combination with PD0325901, A 83-01 also enables OCT4-mediated reprogramming (Zhu et al. Cell Stem Cell, 2010). A 83-01 is supplied as a crystalline solid with ≥ 95% purity; Molecular weight 421.5 g/mol; CAS number 909910-43-6. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5873 Supplier STEMCELL Technologies Supplier No. 72024
Inhibit activin receptor-like kinase (ALK), including ALK5 (type I transforming growth factor-β receptor), ALK4 (type IB activin receptor), and ALK7 (type I NODAL receptor) with A 83-01 (Tojo et al. Cancer Sci, 2005). A 83-01 inhibits mesoderm and endoderm specification (Zhang et al. Cell Stem Cell, 2016), facilitates the conversion of epiblast stem cells to the naїve pluripotent state (Zhou et al. J Biol Chem, 2010; Li et al. Cell Stem Cell, 2009), and disrupts epithelial to mesenchymal transition through inhibition of the SMAD signaling pathway (Tojo et al. Cancer Sci, 2005). In combination with PD0325901, A 83-01 also enables OCT4-mediated reprogramming (Zhu et al. Cell Stem Cell, 2010). A 83-01 is supplied as a crystalline solid with ≥ 95% purity; Molecular weight 421.5 g/mol; CAS number 909910-43-6. Visit STEMCELL.com for more information and related products.